Methyl 5-bromo-6-fluoropicolinate 98%

Supplier: Ambeed
Warning

A1575070-5G A1575070-1G A1575070-250MG A1575070-25G
77567-290EA 259 USD
77567-290 77598-074 77876-319 77876-320
Methyl 5-bromo-6-fluoropicolinate 98%
Methyl 5-bromo-6-fluoropicolinate

Precaución: For research use only.

Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
MDL Number: MFCD14698116
CAS Number: 1214336-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR