DMT-2′O-MOE-rMeU phosphoramidite 95%

Supplier: Ambeed
Warning

A1538640-1G A1538640-250MG A1538640-100MG A1538640-5G
77567-238EA 63 USD
77567-238 77567-240 77597-994 77597-996
DMT-2′O-MOE-rMeU phosphoramidite 95%
DMT-2′O-MOE-rMeU phosphoramidite

Precaución: For research use only.

Formula: C₄₃H₅₅N₄O₁₀P
MDL Number: MFCD11505865
CAS Number: 163878-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR