axial (E)-Cyclooct-2-enol 98%

Supplier: Ambeed
Danger

A1535951-250MG A1535951-100MG
77567-232EA 379.04 USD
77567-232 77625-322
axial (E)-Cyclooct-2-enol 98%
axial (E)-Cyclooct-2-enol

Precaución: For research use only.

Formula: C₈H₁₄O
MW: 126.196 g/mol
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR