6-Chloro-5-fluoropyridin-3-amine 98%

Supplier: Ambeed
Warning

A152835-1G A152835-100MG A152835-250MG A152835-5G
77567-216EA 120 USD
77567-216 77883-196 77883-197 77883-198
6-Chloro-5-fluoropyridin-3-amine 98%
6-Chloro-5-fluoropyridin-3-amine

Precaución: For research use only.

Formula: C₅H₄ClFN₂
MDL Number: MFCD18257897
CAS Number: 1256806-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR