(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Hse(Me)-OH

A152750-5G A152750-250MG
77567-212EA 405.82 USD
77567-212 77611-438
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybutanoic acid

Precaución: For research use only.

Formula: C₂₀H₂₁NO₅
MW: 355.38 g/mol
MDL Number: MFCD00270543
CAS Number: 173212-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR