(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 98%

Supplier: Ambeed
Warning

Synonyms: rel-(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol, BCN-OH

A1523159-1G A1523159-100MG A1523159-50MG A1523159-250MG A1523159-5G
77567-200EA 387.28 USD
77567-200 77600-170 77597-940 77625-240 77625-242
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 98%
(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol

Precaución: For research use only.

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD26142970
CAS Number: 1263166-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR