tert-Butyl 2-(4-hydroxypiperidin-4-yl)acetate 98%

Supplier: Ambeed
Warning

A1510297-1G A1510297-250MG A1510297-100MG
77567-178EA 25 USD
77567-178 77597-848 77624-808
tert-Butyl 2-(4-hydroxypiperidin-4-yl)acetate 98%
tert-Butyl 2-(4-hydroxypiperidin-4-yl)acetate

Precaución: For research use only.

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD24565493
CAS Number: 180532-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR