Tri-GalNAc(OAc)3 TFA 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-11,17-dioxo-6-[[3-oxo-3-[[3-[[1-oxo-5-[[3,4,6-tri-O-acetyl-2-(acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]pentyl]amino]propyl]amino]propoxy]methyl]-N-[3-[[1-oxo-5-[[3,4,6-tri-O-acetyl-2-(acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]pentyl]amino]propyl]-21-[[3,4,6-tri-O-acetyl-2-(acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]-4,8-dioxa-12,16-diazaheneicosanamide 2,2,2-trifluoroacetate

A1504298-100MG A1504298-50MG
77567-160EA 523 USD
77567-160 77600-130
Tri-GalNAc(OAc)3 TFA 98%
Tri-GalNAc(OAc)3 TFA

Precaución: For research use only.

Formula: C₈₁H₁₂₉F₃N₁₀O₃₈
CAS Number: 1159408-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR