2,4,6-Trifluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A1503781-5G A1503781-100G
77567-156EA 26 USD
77567-156 77875-838
2,4,6-Trifluorobenzaldehyde 98%
2,4,6-Trifluorobenzaldehyde

Precaución: For research use only.

Formula: C₇H₃F₃O
MDL Number: MFCD00061197
CAS Number: 58551-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR