5,7-Di-tert-butyl-3-(3,5-dimethylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1503656-1G A1503656-100MG A1503656-250MG A1503656-5G
77567-154EA 241 USD
77567-154 77597-790 77597-792 77875-834
5,7-Di-tert-butyl-3-(3,5-dimethylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-(3,5-dimethylphenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Precaución: For research use only.

Formula: C₂₃H₃₀BF₄NO
CAS Number: 2750161-98-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR