Bis(4-chlorophenyl)iodonium trifluoromethanesulfonate 97%

Supplier: Ambeed
Danger

A1497692-5G A1497692-25G
77567-138EA 162 USD
77567-138 77875-682
Bis(4-chlorophenyl)iodonium trifluoromethanesulfonate 97%
Bis(4-chlorophenyl)iodonium trifluoromethanesulfonate

Precaución: For research use only.

Formula: C₁₃H₈Cl₂F₃IO₃S
CAS Number: 175391-06-7
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR