tert-Butyl ((S)-5-((S)-2-amino-3-methylbutanamido)-6-((4-(hydroxymethyl)phenyl)amino)-6-oxohexyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: PAB-Val-Lys-Boc

A1497380-1G A1497380-100MG A1497380-250MG
77567-134EA 657.14 USD
77567-134 77624-610 77624-612
tert-Butyl ((S)-5-((S)-2-amino-3-methylbutanamido)-6-((4-(hydroxymethyl)phenyl)amino)-6-oxohexyl)carbamate 97%
tert-Butyl ((S)-5-((S)-2-amino-3-methylbutanamido)-6-((4-(hydroxymethyl)phenyl)amino)-6-oxohexyl)carbamate

Precaución: For research use only.

Formula: C₂₃H₃₈N₄O₅
MW: 450.57 g/mol
MDL Number: MFCD34187221
CAS Number: 1432969-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR