5-(Diphenylphosphanyl)-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A1488816-1G A1488816-250MG A1488816-100MG
77567-102EA 393.46 USD
77567-102 77597-640 77624-456
5-(Diphenylphosphanyl)-2-(trifluoromethyl)pyridine 98%
5-(Diphenylphosphanyl)-2-(trifluoromethyl)pyridine

Precaución: For research use only.

Formula: C₁₈H₁₃F₃NP
MW: 331.27 g/mol
MDL Number: MFCD34563796
CAS Number: 2408844-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR