2-(Methylamino)pyridine 98+%

Supplier: Ambeed
Warning

A1488251-25G A1488251-5G
77567-098EA 57.68 USD
77567-098 77624-454
2-(Methylamino)pyridine 98+%
2-(Methylamino)pyridine

Precaución: For research use only.

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00006250
CAS Number: 4597-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR