(S)-3-Bromo-2-(1-methoxyethyl)pyridine 97%

Supplier: Ambeed
Warning

A1476254-1G A1476254-25G
77567-050EA 36.05 USD
77567-050 77597-498
(S)-3-Bromo-2-(1-methoxyethyl)pyridine 97%
(S)-3-Bromo-2-(1-methoxyethyl)pyridine

Precaución: For research use only.

Formula: C₈H₁₀BrNO
MW: 216.0751 g/mol
CAS Number: 2641451-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR