(2S,4R)-1-((S)-2-(11-Aminoundecanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-Me-C10-NH2

A1471141-250MG A1471141-50MG
77567-026EA 306.94 USD
77567-026 77622-222
(2S,4R)-1-((S)-2-(11-Aminoundecanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide 95%
(2S,4R)-1-((S)-2-(11-Aminoundecanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide

Precaución: For research use only.

Formula: C₃₄H₅₃N₅O₄S
MW: 627.8807 g/mol
MDL Number: MFCD33029233
CAS Number: 2376139-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR