m-PEG-thiol, M.W. ~ 2000

Supplier: Ambeed
Danger

A1470854-1G A1470854-250MG A1470854-5G
77567-020EA 63 USD
77567-020 77875-064 77875-065
m-PEG-thiol, M.W. ~ 2000
m-PEG-thiol

Precaución: For research use only.

MDL Number: MFCD21363241
CAS Number: 134874-49-0
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR