4-Hydroxy-4-methylcyclohexanone 95%

Supplier: Ambeed
Warning

A147051-1G A147051-100MG A147051-250MG
77567-018EA 283 USD
77567-018 77881-146 77881-147
4-Hydroxy-4-methylcyclohexanone 95%
4-Hydroxy-4-methylcyclohexanone

Precaución: For research use only.

Formula: C₇H₁₂O₂
MW: 128.169 g/mol
MDL Number: MFCD13659404
CAS Number: 17429-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR