N-(Adamantan-1-yl)-1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilan-2-amine 97%

Supplier: Ambeed
Danger

A1469490-1G A1469490-5G
77567-010EA 351.23 USD
77567-010 77567-012
N-(Adamantan-1-yl)-1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilan-2-amine 97%
N-(Adamantan-1-yl)-1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilan-2-amine

Precaución: For research use only.

Formula: C₁₉H₄₃NSi₄
MW: 397.89 g/mol
MDL Number: MFCD32857305
CAS Number: 2451224-01-2
UN: 2925
ADR: 4.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR