L-Cysteine S-sulfate sodium salt sesquihydrate 98%

Supplier: Ambeed
Warning

Synonyms: H-Cys(SO₃Na)-OH·3/2H₂O

A1466098-100G A1466098-1G A1466098-5G
77566-992EA 396 USD
77566-992 77597-372 77597-374
L-Cysteine S-sulfate sodium salt sesquihydrate 98%
L-Cysteine S-sulfate sodium salt sesquihydrate

Precaución: For research use only.

Formula: C₆H₁₈N₂Na₂O₁₃S₄
MW: 500.4523 g/mol
MDL Number: MFCD32669873
CAS Number: 150465-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR