Hyaluronic acid sodium salt, M.W. 1500000-1750000

Supplier: Ambeed
Warning

A1463896-5G A1463896-25G
77566-982EA 36 USD
77566-982 77597-340
Hyaluronic acid sodium salt, M.W. 1500000-1750000
Hyaluronic acid sodium salt

Precaución: For research use only.

Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR