Hyaluronic acid sodium salt, M.W. 130000-150000

Supplier: Ambeed
Warning

Synonyms: Sodium hyaluronate, Hyaluronic acid sodium, HA sodium salt

A1463894-25G A1463894-100G A1463894-5G
77566-980EA 45.32 USD
77566-980 77597-336 77597-338
Hyaluronic acid sodium salt, M.W. 130000-150000
Hyaluronic acid sodium salt

Precaución: For research use only.

Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR