1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 98%

Supplier: Ambeed
Danger

A1463109-250MG A1463109-100MG
77566-978EA 168 USD
77566-978 77597-324
1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 98%
1,3-Bis((R)-1-(naphthalen-1-yl)ethyl)-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate

Precaución: For research use only.

Formula: C₂₇H₂₇BF₄N₂
MDL Number: MFCD32197455
CAS Number: 1344155-18-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR