5-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%

Supplier: Ambeed
Warning

A146040-25G A146040-100G A146040-500G
77566-970EA 17 USD
77566-970 77876-991 77876-992
5-Amino-1,3-dihydro-2H-benzimidazol-2-one 98%
5-Amino-1,3-dihydro-2H-benzimidazol-2-one

Precaución: For research use only.

Formula: C₇H₇N₃O
MDL Number: MFCD00053555
CAS Number: 95-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR