D-Panthenol 98%

Supplier: Ambeed
Warning

A1459790-25G A1459790-1000G
77566-968EA 8 USD
77566-968 77874-958
D-Panthenol 98%
D-Panthenol

Precaución: For research use only.

Formula: C₉H₁₉NO₄
MDL Number: MFCD00065006
CAS Number: 81-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR