Ethylene-vinyl acetate copolymer vinyl acetate 40% (w/w) melt index ((52 g/10 min (190°C/2.16kg))

Supplier: Ambeed
Warning

A1456035-25G A1456035-1000G A1456035-100G
77566-952EA 6 USD
77566-952 77874-935 77874-936
Ethylene-vinyl acetate copolymer vinyl acetate 40% (w/w) melt index ((52 g/10 min (190°C/2.16kg))
Ethylene-vinyl acetate copolymer

Precaución: For research use only.

Formula: (CH₂CH₂)m[CH₂CH(OCOCH₃)]n
MDL Number: MFCD00133996
CAS Number: 24937-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR