3-Bromo-5-chloro-4-fluorophenol 97%

Supplier: Ambeed
Warning

A145255-1G A145255-5G
77566-930EA 391 USD
77566-930 77900-823
3-Bromo-5-chloro-4-fluorophenol 97%
3-Bromo-5-chloro-4-fluorophenol

Precaución: For research use only.

Formula: C₆H₃BrClFO
MDL Number: MFCD28166393
CAS Number: 1805518-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR