N-(trans-4-(3-Chloro-4-cyanophenoxy)cyclohexyl)-6-(4-((4-(2-(2,6-dioxopiperidin-3-yl)-6-fluoro-1,3-dioxoisoindolin-5-yl)piperazin-1-yl)methyl)piperidin-1-yl)pyridazine-3-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: Bavdegalutamide

A1452141-250MG A1452141-100MG A1452141-5G
77566-926EA 161 USD
77566-926 77597-226 77874-916
N-(trans-4-(3-Chloro-4-cyanophenoxy)cyclohexyl)-6-(4-((4-(2-(2,6-dioxopiperidin-3-yl)-6-fluoro-1,3-dioxoisoindolin-5-yl)piperazin-1-yl)methyl)piperidin-1-yl)pyridazine-3-carboxamide 97%
N-(trans-4-(3-Chloro-4-cyanophenoxy)cyclohexyl)-6-(4-((4-(2-(2,6-dioxopiperidin-3-yl)-6-fluoro-1,3-dioxoisoindolin-5-yl)piperazin-1-yl)methyl)piperidin-1-yl)pyridazine-3-carboxamide

Precaución: For research use only.

Formula: C₄₁H₄₃ClFN₉O₆
CAS Number: 2222112-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR