(2,2'-Bipyridine-κN1,κN1')tris(trifluoromethyl)-Copper 98%

Supplier: Ambeed
Danger

A1445424-1G A1445424-100MG A1445424-250MG A1445424-5G
77566-922EA 325.48 USD
77566-922 77597-208 77597-210 77597-212
(2,2'-Bipyridine-κN1,κN1')tris(trifluoromethyl)-Copper 98%
(2,2'-Bipyridine-κN1,κN1')tris(trifluoromethyl)-Copper

Precaución: For research use only.

Formula: C₁₃H₈CuF₉N₂
MW: 426.75 g/mol
CAS Number: 1680206-12-5
UN: 1479
ADR: 5.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR