2-Bromo-7-chlorodibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1442004-5G A1442004-1G
77566-902EA 169.95 USD
77566-902 77622-092
2-Bromo-7-chlorodibenzo[b,d]furan 97%
2-Bromo-7-chlorodibenzo[b,d]furan

Precaución: For research use only.

Formula: C₁₂H₆BrClO
MW: 281.53 g/mol
MDL Number: MFCD34476115
CAS Number: 2355229-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR