tert-Butyl N-(3-iodopropyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-iodopropyl)carbamate

A143903-1G A143903-5G A143903-250MG A143903-25G
77566-890EA 89 USD
77566-890 77566-892 77600-074 77871-701
tert-Butyl N-(3-iodopropyl)carbamate 95%
tert-Butyl N-(3-iodopropyl)carbamate

Precaución: For research use only.

Formula: C₈H₁₆INO₂
MW: 285.123 g/mol
MDL Number: MFCD09951819
CAS Number: 167479-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR