Isostearyl alcohol 98+%

Supplier: Ambeed
Warning

A143651-500G A143651-25G A143651-1000G A143651-100G
77566-880EA 131 USD
77566-880 77879-009 77879-007 77879-008
Isostearyl alcohol 98+%
Isostearyl alcohol

Precaución: For research use only.

Formula: C₁₈H₃₈O
CAS Number: 27458-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR