(3-Bromonaphthalen-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A1420906-5G A1420906-1G A1420906-250MG
77566-858EA 264 USD
77566-858 77874-778 77874-779
(3-Bromonaphthalen-2-yl)boronic acid 98%
(3-Bromonaphthalen-2-yl)boronic acid

Precaución: For research use only.

Formula: C₁₀H₈BBrO₂
MDL Number: MFCD18410706
CAS Number: 1301205-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR