5-(Pyridin-2-yldisulfanyl)pentanoic acid 98%

Supplier: Ambeed
Warning

A1412232-1G A1412232-100MG A1412232-250MG
77566-846EA 145.23 USD
77566-846 77597-164 77597-166
5-(Pyridin-2-yldisulfanyl)pentanoic acid 98%
5-(Pyridin-2-yldisulfanyl)pentanoic acid

Precaución: For research use only.

Formula: C₁₀H₁₃NO₂S₂
MW: 243.3457 g/mol
MDL Number: MFCD27935400
CAS Number: 250266-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR