(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(oxazol-5-yl)propanoic acid 95%

Supplier: Ambeed
Warning

A140734-1G A140734-100MG A140734-250MG
77566-836EA 679 USD
77566-836 77891-979 77891-980
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(oxazol-5-yl)propanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(oxazol-5-yl)propanoic acid

Precaución: For research use only.

Formula: C₂₁H₁₈N₂O₅
MDL Number: MFCD28134507
CAS Number: 1956434-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR