Nickel(II) tetrafluoroborate hexahydrate 98%

Supplier: Ambeed
Danger

A140210-25G A140210-100G A140210-500G
77566-816EA 34 USD
77566-816 77878-987 77878-988
Nickel(II) tetrafluoroborate hexahydrate 98%
Nickel(II) tetrafluoroborate hexahydrate

Precaución: For research use only.

Formula: B₂F₈Ni·6H₂O
MW: 340.394 g/mol
MDL Number: MFCD00150259
CAS Number: 15684-36-3
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR