(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9-trioxo-2-oxa-4,7,10-triazadodecan-12-oic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Gly-Gly-Phe-OH

A1385147-5G A1385147-250MG
77566-784EA 128.75 USD
77566-784 77597-100
(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9-trioxo-2-oxa-4,7,10-triazadodecan-12-oic acid 98%
(S)-11-Benzyl-1-(9H-fluoren-9-yl)-3,6,9-trioxo-2-oxa-4,7,10-triazadodecan-12-oic acid

Precaución: For research use only.

Formula: C₂₈H₂₇N₃O₆
MW: 501.531 g/mol
MDL Number: MFCD33023222
CAS Number: 160036-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR