2-(8-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1375150-5G A1375150-1G A1375150-250MG A1375150-100MG
77566-748EA 196 USD
77566-748 77592-612 77592-614 77867-708
2-(8-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(8-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₆H₁₈BClO₂
MDL Number: MFCD32689659
CAS Number: 2454397-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR