7-Fluoronaphthalene-1,3-diol 98%
Supplier: Ambeed
|
Danger
|
A1374994-25G
A1374994-5G
A1374994-1G
77566-746EA
67
USD
77566-746
77592-610
77621-940
7-Fluoronaphthalene-1,3-diol 98%
7-Fluoronaphthalene-1,3-diol
Precaución: For research use only.
Formula:
C₁₀H₇FO₂ |
MDL Number:
MFCD33022268 CAS Number: 2460027-79-4 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...