7-Bromo-4,4-difluoro-3,4-dihydronaphthalen-1(2H)-one 95%

Supplier: Ambeed
Warning

A136683-1G A136683-250MG
77566-716EA 1097 USD
77566-716 77685-178
7-Bromo-4,4-difluoro-3,4-dihydronaphthalen-1(2H)-one 95%
7-Bromo-4,4-difluoro-3,4-dihydronaphthalen-1(2H)-one

Precaución: For research use only.

Formula: C₁₀H₇BrF₂O
MDL Number: MFCD23703392
CAS Number: 1415108-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR