[5-[Bis(1,1-dimethylethyl)phosphino-κP]-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole][(1,2,3-η)-1-phenyl-2-propen-1-yl]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) 97%

Supplier: Ambeed
Danger

A1360929-250MG A1360929-100MG A1360929-1G
77566-696EA 427 USD
77566-696 77621-748 77867-546
[5-[Bis(1,1-dimethylethyl)phosphino-κP]-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole][(1,2,3-η)-1-phenyl-2-propen-1-yl]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1) 97%
[5-[Bis(1,1-dimethylethyl)phosphino-κP]-1',3',5'-triphenyl-1,4'-bi-1H-pyrazole][(1,2,3-η)-1-phenyl-2-propen-1-yl]-Palladium(1+), 1,1,1-trifluoromethanesulfonate (1:1)

Precaución: For research use only.

Formula: C₄₂H₄₄F₃N₄O₃PPdS
MW: 879.278 g/mol
CAS Number: 1846558-62-0
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR