Methoxypolyethylene glycol amine, M.W. 5000

Supplier: Ambeed
Warning

A1360369-1G A1360369-250MG
77566-694EA 76 USD
77566-694 77621-730
Methoxypolyethylene glycol amine, M.W. 5000
Methoxypolyethylene glycol amine

Precaución: For research use only.

Formula: H₂NCH₂CH₂(OCH₂CH₂)nOCH₃
MDL Number: MFCD00131599
CAS Number: 80506-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR