2-Methylthiazole-5-boronic acid, pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole

A1359088-1G A1359088-250MG A1359088-100MG A1359088-25G
77566-686EA 77 USD
77566-686 77566-688 77621-712 77867-509
2-Methylthiazole-5-boronic acid, pinacol ester 96%
2-Methylthiazole-5-boronic acid, pinacol ester

Precaución: For research use only.

Formula: C₁₀H₁₆BNO₂S
MDL Number: MFCD10697442
CAS Number: 1218791-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR