(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine oxide) 98%

Supplier: Ambeed
Warning

A1357030-1G A1357030-100MG A1357030-250MG A1357030-5G
77566-672EA 405.82 USD
77566-672 77596-870 77596-872 77621-708
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine oxide) 98%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine oxide)

Precaución: For research use only.

Formula: C₃₉H₃₂O₃P₂
MW: 610.6171 g/mol
MDL Number: MFCD32670680
CAS Number: 808142-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR