3-(5-Bromo-3-methyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidine-2,6-dione 98%

Supplier: Ambeed
Warning

A1355806-1G A1355806-250MG
77566-664EA 211.15 USD
77566-664 77621-698
3-(5-Bromo-3-methyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidine-2,6-dione 98%
3-(5-Bromo-3-methyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidine-2,6-dione

Precaución: For research use only.

Formula: C₁₃H₁₂BrN₃O₃
MW: 338.157 g/mol
MDL Number: MFCD34595287
CAS Number: 2300099-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR