11bS-N,N-Bis[(1R)-1-(1-naphthalenyl)ethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine 95%

Supplier: Ambeed
Warning

A1354020-100MG A1354020-250MG A1354020-1G
77566-652EA 70 USD
77566-652 77621-692 77867-475
11bS-N,N-Bis[(1R)-1-(1-naphthalenyl)ethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine 95%
11bS-N,N-Bis[(1R)-1-(1-naphthalenyl)ethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

Precaución: For research use only.

Formula: C₄₄H₃₄NO₂P
MDL Number: MFCD11973798
CAS Number: 342813-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR