2-Fluoro-5-iodophenol 97%

Supplier: Ambeed
Warning

A134570-1G A134570-250MG
77566-636EA 60 USD
77566-636 77871-640
2-Fluoro-5-iodophenol 97%
2-Fluoro-5-iodophenol

Precaución: For research use only.

Formula: C₆H₄FIO
MDL Number: MFCD16659616
CAS Number: 186589-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR