2-Chloro-3-methoxy-5-(trifluoromethyl)pyridine 95%
Supplier: Ambeed
|
Warning
|
A134250-1G
A134250-100MG
77566-632EA
350
USD
77566-632
77885-154
2-Chloro-3-methoxy-5-(trifluoromethyl)pyridine 95%
2-Chloro-3-methoxy-5-(trifluoromethyl)pyridine
Precaución: For research use only.
Formula:
C₇H₅ClF₃NO |
MDL Number:
MFCD16610622 CAS Number: 1227563-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...