1,5-Dichloropentane-2,4-dione 97%

Supplier: Ambeed
Danger

A1340600-100MG A1340600-250MG
77566-626EA 172 USD
77566-626 77867-401
1,5-Dichloropentane-2,4-dione 97%
1,5-Dichloropentane-2,4-dione

Precaución: For research use only.

Formula: C₅H₆Cl₂O₂
MDL Number: MFCD07357286
CAS Number: 40630-12-4
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR