2-(Difluoromethoxy)benzaldehyde 96%

Supplier: Ambeed
Warning

A134044-1G A134044-10G A134044-5G
77566-622EA 13 USD
77566-622 77592-546 77685-159
2-(Difluoromethoxy)benzaldehyde 96%
2-(Difluoromethoxy)benzaldehyde

Precaución: For research use only.

Formula: C₈H₆F₂O₂
MDL Number: MFCD00042251
CAS Number: 71653-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR